Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY6G5C
Entrez Gene ID
102117849
Gene Name
lymphocyte antigen 6 family member G5C
Gene Aliases
-
Chromosome Location
Chr.4: 139191218 - 139195264 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553490.2 XP_005553547.1 1 - 2 481 92

View Full Product Details