Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACRT
Entrez Gene ID
102118200
Gene Name
lacritin
Gene Aliases
-
Chromosome Location
Chr.11: 53561907 - 53566036 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571097.2 XP_005571154.1 3 - 4 298 81
GenBank mRNA DQ659149.1 - 3 - 4 250 81

View Full Product Details