Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNLT1
Entrez Gene ID
102145906
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
-
Chromosome Location
Chr.4: 65050789 - 65058795 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552217.2 XP_005552274.1 3 - 4 270 65

View Full Product Details