Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCPIP1
Entrez Gene ID
102142674
Gene Name
valosin containing protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 67869443 - 67911095 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563455.2 XP_005563512.1 2 - 3 3818 77
GenBank mRNA AB056821.1 - 3 - 4 983 77

View Full Product Details