The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005547524.2 | XP_005547581.1 | 5 - 6 | 1008 | 69 |
XM_005547525.2 | XP_005547582.1 | 4 - 5 | 970 | 69 |