Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM92
Entrez Gene ID
102130289
Gene Name
transmembrane protein 92
Gene Aliases
-
Chromosome Location
Chr.16: 35670403 - 35677425 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583662.2 XP_005583719.1 2 - 3 167 69

View Full Product Details