Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYRM4
Entrez Gene ID
102128026
Gene Name
LYR motif containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 165723787 - 165878088 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554126.2 XP_005554183.1 2 - 3 424 99

View Full Product Details