Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2E2
Entrez Gene ID
102138693
Gene Name
ubiquitin conjugating enzyme E2 E2
Gene Aliases
-
Chromosome Location
Chr.2: 23903136 - 24287142 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545656.2 XP_005545713.1 1 - 2 203 74

View Full Product Details