Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL6IP6
Entrez Gene ID
102120116
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 6
Gene Aliases
-
Chromosome Location
Chr.12: 41465544 - 41509113 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573195.2 XP_005573252.1 1 - 2 1138 134

View Full Product Details