Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H4orf27
Entrez Gene ID
102117500
Gene Name
chromosome 5 open reading frame, human C4orf27
Gene Aliases
-
Chromosome Location
Chr.5: 168939462 - 168970016 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556289.2 XP_005556346.1 2 - 3 285 149
XM_005556290.2 XP_005556347.1 2 - 3 286 149
XM_005556291.2 XP_005556348.1 2 - 3 285 149

View Full Product Details