Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZG16
Entrez Gene ID
102123774
Gene Name
zymogen granule protein 16
Gene Aliases
-
Chromosome Location
Chr.20: 28234660 - 28236883 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591577.1 XP_005591634.1 3 - 4 256 64

View Full Product Details