Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JCHAIN
Entrez Gene ID
102144919
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGJ
Chromosome Location
Chr.5: 63920652 - 63932185 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555133.1 XP_005555190.1 1 - 2 246 80

View Full Product Details