Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM89A
Entrez Gene ID
102143299
Gene Name
family with sequence similarity 89 member A
Gene Aliases
-
Chromosome Location
Chr.1: 18524813 - 18543588 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005539790.2 XP_005539847.1 1 - 2 334 69

View Full Product Details