Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLOC1S2
Entrez Gene ID
102120191
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
-
Chromosome Location
Chr.9: 99185327 - 99195140 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287735.1 NP_001274664.1 3 - 4 313 134
XM_015455892.1 XP_015311378.1 3 - 4 605 134
XM_015455893.1 XP_015311379.1 2 - 3 890 134
GenBank mRNA AB168891.1 - 3 - 4 313 134

View Full Product Details