Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGN
Entrez Gene ID
102115841
Gene Name
serglycin
Gene Aliases
-
Chromosome Location
Chr.9: 66599167 - 66614195 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565677.2 XP_005565734.1 2 - 3 308 70

View Full Product Details