Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OOEP
Entrez Gene ID
102134332
Gene Name
oocyte expressed protein
Gene Aliases
-
Chromosome Location
Chr.4: 100555994 - 100557947 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552558.2 XP_005552615.1 1 - 2 883 96

View Full Product Details