Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAU
Entrez Gene ID
102117052
Gene Name
FAU ubiquitin like and ribosomal protein S30 fusion
Gene Aliases
-
Chromosome Location
Chr.14: 9408574 - 9410188 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577315.2 XP_005577372.1 1 - 2 147 88

View Full Product Details