Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM24
Entrez Gene ID
102126848
Gene Name
small integral membrane protein 24
Gene Aliases
C19H19orf77
Chromosome Location
Chr.19: 3442194 - 3447947 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587501.2 XP_005587558.2 3 - 4 311 81

View Full Product Details