Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102146847
Entrez Gene ID
102146847
Gene Name
guanylate-binding protein 3
Gene Aliases
GBP3
Chromosome Location
Chr.1: 138227583 - 138243741 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542800.2 XP_005542857.1 2 - 3 1001 157

View Full Product Details