Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRI3BP
Entrez Gene ID
102144651
Gene Name
BRI3 binding protein
Gene Aliases
-
Chromosome Location
Chr.11: 129371502 - 129405439 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572605.1 XP_005572662.1 2 - 3 401 64

View Full Product Details