Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB20
Entrez Gene ID
102130428
Gene Name
RAB20, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.17: 92929457 - 92966583 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586238.2 XP_005586295.1 1 - 2 449 80

View Full Product Details