Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX4
Entrez Gene ID
102135462
Gene Name
ALX homeobox 4
Gene Aliases
-
Chromosome Location
Chr.14: 23277750 - 23324596 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578128.2 XP_005578185.1 3 - 4 1081 118

View Full Product Details