Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK12
Entrez Gene ID
102146069
Gene Name
potassium two pore domain channel subfamily K member 12
Gene Aliases
-
Chromosome Location
Chr.13: 62806383 - 62861031 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575943.2 XP_005576000.1 1 - 2 420 124

View Full Product Details