Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC8E
Entrez Gene ID
102129159
Gene Name
leucine rich repeat containing 8 VRAC subunit E
Gene Aliases
-
Chromosome Location
Chr.19: 8419118 - 8433372 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587811.2 XP_005587868.1 1 - 2 92 117

View Full Product Details