Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR7
Entrez Gene ID
102127418
Gene Name
proline rich 7, synaptic
Gene Aliases
-
Chromosome Location
Chr.6: 177273121 - 177283329 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558648.2 XP_005558705.1 2 - 3 1678 72

View Full Product Details