Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCTD12
Entrez Gene ID
102124540
Gene Name
potassium channel tetramerization domain containing 12
Gene Aliases
-
Chromosome Location
Chr.17: 59389056 - 59395277 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586032.2 XP_005586089.1 1 - 1 5826 158
GenBank mRNA AB173887.1 - 1 - 1 158

View Full Product Details