Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX2-3
Entrez Gene ID
102140990
Gene Name
NK2 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.9: 98400551 - 98404249 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566164.1 XP_005566221.1 1 - 2 569 148

View Full Product Details