Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS12
Entrez Gene ID
102131525
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
-
Chromosome Location
Chr.5: 121717388 - 121736099 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555866.2 XP_005555923.1 1 - 2 263 78

View Full Product Details