Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGN
Entrez Gene ID
102119950
Gene Name
gametogenetin
Gene Aliases
-
Chromosome Location
Chr.19: 38833162 - 38837548 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015441757.1 XP_015297243.1 3 - 4 2267 99

View Full Product Details