Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPRV1
Entrez Gene ID
102122540
Gene Name
aspartic peptidase retroviral like 1
Gene Aliases
-
Chromosome Location
Chr.13: 39976633 - 39978811 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575666.2 XP_005575723.1 1 - 1 450 124

View Full Product Details