Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG3
Entrez Gene ID
102116655
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
Gene Aliases
-
Chromosome Location
Chr.13: 67948733 - 68006090 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576015.2 XP_005576072.1 1 - 2 664 63

View Full Product Details