Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAMEF12
Entrez Gene ID
102140054
Gene Name
PRAME family member 12
Gene Aliases
-
Chromosome Location
Chr.1: 215292297 - 215295114 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544754.2 XP_005544811.1 2 - 3 877 135

View Full Product Details