Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD8
Entrez Gene ID
102114993
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
Chromosome Location
Chr.5: 44040134 - 44330390 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554790.2 XP_005554847.1 1 - 2 3269 137

View Full Product Details