Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SVBP
Entrez Gene ID
102116561
Gene Name
small vasohibin binding protein
Gene Aliases
CCDC23
Chromosome Location
Chr.1: 184867282 - 184879715 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543713.2 XP_005543770.1 2 - 3 260 106

View Full Product Details