Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD18
Entrez Gene ID
102145937
Gene Name
abhydrolase domain containing 18
Gene Aliases
C5H4orf29
Chromosome Location
Chr.5: 126915764 - 127005351 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015450814.1 XP_015306300.1 4 - 5 633 114
XM_015450815.1 XP_015306301.1 3 - 4 521 114
XM_015450816.1 XP_015306302.1 3 - 4 1027 114
XM_015450817.1 XP_015306303.1 4 - 5 633 114
XM_015450818.1 XP_015306304.1 4 - 5 633 114
XM_015450819.1 XP_015306305.1 4 - 5 633 114

View Full Product Details