Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGRP
Entrez Gene ID
102144154
Gene Name
agouti related neuropeptide
Gene Aliases
-
Chromosome Location
Chr.20: 55643376 - 55644902 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592267.2 XP_005592324.1 1 - 2 603 140

View Full Product Details