Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3B
Entrez Gene ID
102122066
Gene Name
RAB3B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 175537216 - 175615683 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543380.2 XP_005543437.1 2 - 3 455 89

View Full Product Details