Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBLN1
Entrez Gene ID
102118708
Gene Name
cerebellin 1 precursor
Gene Aliases
-
Chromosome Location
Chr.20: 37039624 - 37043456 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591869.1 XP_005591926.1 1 - 2 611 83

View Full Product Details