Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX3
Entrez Gene ID
102118621
Gene Name
pannexin 3
Gene Aliases
-
Chromosome Location
Chr.14: 120409123 - 120417172 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580026.2 XP_005580083.1 1 - 2 317 77

View Full Product Details