Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF20
Entrez Gene ID
102141394
Gene Name
transcription factor 20
Gene Aliases
-
Chromosome Location
Chr.10: 8776878 - 8905733 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567107.2 XP_005567164.1 3 - 4 5912 80
XM_005567108.2 XP_005567165.1 3 - 4 5912 80
XM_005567109.2 XP_005567166.1 3 - 4 5912 80
XM_015457109.1 XP_015312595.1 3 - 4 5884 80

View Full Product Details