Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRX5
Entrez Gene ID
102122409
Gene Name
iroquois homeobox 5
Gene Aliases
-
Chromosome Location
Chr.20: 42797392 - 42802488 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591937.2 XP_005591994.1 1 - 2 369 104

View Full Product Details