Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRG3
Entrez Gene ID
102141064
Gene Name
proteoglycan 3, pro eosinophil major basic protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 16243561 - 16248296 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577864.2 XP_005577921.1 4 - 5 669 72

View Full Product Details