Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THY1
Entrez Gene ID
102139037
Gene Name
Thy-1 cell surface antigen
Gene Aliases
-
Chromosome Location
Chr.14: 115449800 - 115456001 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579919.2 XP_005579976.1 2 - 3 1051 117

View Full Product Details