Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BATF
Entrez Gene ID
102144969
Gene Name
basic leucine zipper ATF-like transcription factor
Gene Aliases
-
Chromosome Location
Chr.7: 140235751 - 140260110 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561820.2 XP_005561877.1 1 - 2 317 82

View Full Product Details