Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102124550
Entrez Gene ID
102124550
Gene Name
myosin regulatory light chain 12A
Gene Aliases
MYL12A
Chromosome Location
Chr.18: 64445759 - 64454247 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587102.2 XP_005587159.1 1 - 2 384 154

View Full Product Details