Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2D4
Entrez Gene ID
102133514
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases
-
Chromosome Location
Chr.3: 76478402 - 76508877 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549710.2 XP_005549767.1 1 - 2 79 72
XM_005549711.2 XP_005549768.1 2 - 3 107 72
XM_015447486.1 XP_015302972.1 2 - 3 98 72
XM_015447488.1 XP_015302974.1 1 - 2 96 72

View Full Product Details