Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRX2
Entrez Gene ID
102137905
Gene Name
paired related homeobox 2
Gene Aliases
-
Chromosome Location
Chr.15: 9380948 - 9384638 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015436348.1 XP_015291834.1 2 - 3 367 84

View Full Product Details