Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
102124479
Gene Name
family with sequence similarity 53 member C
Gene Aliases
-
Chromosome Location
Chr.6: 137435444 - 137448638 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557894.2 XP_005557951.1 4 - 5 1102 100
XM_005557895.2 XP_005557952.1 3 - 4 974 100
XM_005557896.2 XP_005557953.1 3 - 4 739 100
XM_015451938.1 XP_015307424.1 3 - 4 308 100

View Full Product Details