Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN12
Entrez Gene ID
102119022
Gene Name
schlafen family member 12
Gene Aliases
-
Chromosome Location
Chr.16: 31773350 - 31803560 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583462.2 XP_005583519.1 2 - 3 1459 138
XM_015438178.1 XP_015293664.1 3 - 4 1509 138

View Full Product Details