Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
102137636
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C5H4orf21
Chromosome Location
Chr.5: 111888583 - 111974129 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555712.2 XP_005555769.1 10 - 11 3157 142
XM_005555713.2 XP_005555770.1 9 - 10 3073 142
XM_005555714.2 XP_005555771.1 8 - 9 3013 142
XM_005555716.2 XP_005555773.1 9 - 10 3073 142
XM_005555717.2 XP_005555774.1 9 - 10 3073 142

View Full Product Details